Order Entry
Italy
ContactUsLinkComponent
(R*,R*)-(2,8-Bis-trifluoromethyl-quinolin-4-yl)-piperidin-2-yl-methanol hydrochloride
Irritant
(R*,R*)-(2,8-Bis-trifluoromethyl-quinolin-4-yl)-piperidin-2-yl-methanol hydrochloride
Catalog # APOSPC404547-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(R*,R*)-(2,8-Bis-trifluoromethyl-quinolin-4-yl)-piperidin-2-yl-methanol hydrochloride
Catalog # APOSPC404547-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :414.773 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:58560-52-4
Formula:C₁₇H₁₆F₆N₂O·ClH
MDL Number:MFCD08437514