1-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1188264-17-6
:
Formula:C₁₂H₁₂FN₃·ClH
MDL Number:MFCD11109892
:253.703 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together