Order Entry
Italy
ContactUsLinkComponent
 
Irritant
3-Fluoro-5-(piperidin-4-yl)pyridine dihydrochloride 97%
Catalog # APOSPC402103-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Fluoro-5-(piperidin-4-yl)pyridine dihydrochloride 97%
Catalog # APOSPC402103-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :253.144 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1638968-09-8
Formula:C₁₀H₁₃FN₂·2ClH
MDL Number:MFCD29075592