2-Cyano-5-(trifluoromethyl)-3-[4-(trifluoromethyl)phenoxy]pyridine
Catalog # APOSPC401045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:332.201 g/mol
Storage Temperature:Ambient
CAS Number:338759-13-0
Formula:C₁₄H₆F₆N₂O
MDL Number:MFCD00664116