2-Cyano-3-(morpholin-4-yl)-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1449117-57-0
:
Formula:C₁₁H₁₀F₃N₃O
MDL Number:MFCD25970573
:257.212 g/mol
Storage Temperature:Ambient
Frequently Bought Together