Order Entry
Italy
ContactUsLinkComponent
2-(3-Bromo-2-chloro-6-fluorophenyl)-3H-imidazo[4,5-c]pyridine
2-(3-Bromo-2-chloro-6-fluorophenyl)-3H-imidazo[4,5-c]pyridine
Catalog # APOSPC400570-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Bromo-2-chloro-6-fluorophenyl)-3H-imidazo[4,5-c]pyridine
Catalog # APOSPC400570-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :326.552 g/mol
Storage Temperature:Ambient
CAS Number:1439824-02-8
Formula:C₁₂H₆BrClFN₃
MDL Number:MFCD22690833