Order Entry
Italy
ContactUsLinkComponent
7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione
7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione
Catalog # APOSPC400568-1G
Supplier:  Apollo Scientific
CAS Number:  
7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione
Catalog # APOSPC400568-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.147 g/mol
Storage Temperature:Ambient
CAS Number:1624260-16-7
Formula:C₇H₈F₂N₂O₂
MDL Number:MFCD27501044