
6-Bromo-2-fluoro-3-methoxypyridine
Catalog # APOSPC400567-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.012 g/mol
Melting Pt:58…60 °C
:GHS07
Storage Temperature:Ambient
CAS Number:850142-73-3
Formula:C₆H₅BrFNO
MDL Number:MFCD04112568