Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine
4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine
4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine
APOSPC400510-1G
:
Apollo Scientific
:
4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine
APOSPC400510-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
213.212 g/mol
:
Ambient
:
118121-89-4
:
C₈H₁₂FN₅O
:
MFCD00181489