
1-(5-Bromo-3-methylpyridin-2-yl)-2,2,2-trifluoroethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1448790-49-5
:
Formula:C₈H₅BrF₃NO
MDL Number:MFCD27992315
:268.031 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together