
1-(3-Bromo-5-fluoropyridin-2-yl)-2,2,2-trifluoroethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2231675-78-6
:
Formula:C₇H₂BrF₄NO
MDL Number:MFCD31621151
:271.994 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together