
1-(5-Bromo-3-chloropyridin-2-yl)-2,2,2-trifluoroethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2231673-26-8
:
Formula:CâHâBrClFâNO
MDL Number:MFCD31621135
:288.449 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together