
1-(6-Chloro-5-fluoropyridin-3-yl)-2,2,2-trifluoroethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1245915-43-8
:
Formula:C₇H₂ClF₄NO
MDL Number:MFCD18254428
:227.543 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together