4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)-2-hydroxybenzaldehyde
Catalog # APOSPC400068-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:230.143 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:308085-25-8
Formula:C₉H₅F₃N₂O₂
MDL Number:MFCD30468740