Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one
6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one 95+%
6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one 95+%
APOSPC400050-1G
:
Apollo Scientific
:
6,7-Difluoro-2-mercaptoquinazolin-4(3H)-one 95+%
APOSPC400050-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
214.192 g/mol
:
GHS07
:
Ambient
:
1593461-98-3
:
C₈H₄F₂N₂OS
:
MFCD31561231