

5-(Difluoromethyl)pyridine-2-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1211540-57-6
:
Formula:C₇H₄F₂N₂
MDL Number:MFCD18256046
:154.117 g/mol
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together