Order Entry
Italy
ContactUsLinkComponent
2-(4-(Trifluoromethyl)phenyl)morpholine
2-(4-(Trifluoromethyl)phenyl)morpholine
Catalog # APOSPC400017-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-(Trifluoromethyl)phenyl)morpholine
Catalog # APOSPC400017-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.214 g/mol
Storage Temperature:Ambient
CAS Number:62243-72-5
Formula:C₁₁H₁₂F₃NO
MDL Number:MFCD11646253