
(2S)-(-)-3,3,3-Trifluoro-1,2-propenoxide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:130025-34-2
:
Formula:C₃H₃F₃O
MDL Number:MFCD04128982
:112.051 g/mol
Boiling Pt:25…32 °C
Melting Pt:–80 °C
Density:1.294 g/cm³ (25 °C)
Flash Pt:–26 °C
Storage Temperature:Freezer
UN:1993
ADR:3,I
:GHS02
Frequently Bought Together