
Ethyl 8-nitro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2379918-47-3
:
Formula:C₁₁H₈F₃N₃O₄
MDL Number:MFCD32200423
:303.194 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together