
(1R)-2-Fluoro-1-phenylethanamine, N-BOC protected 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2006287-04-1
:
Formula:C₁₃H₁₈FNO₂
MDL Number:MFCD30378945
:239.286 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together