Order Entry
Italy
ContactUsLinkComponent
4-[(2,6-Dichloro-5-fluoropyridin-3-yl)carbonyl]morpholine
Irritant
4-[(2,6-Dichloro-5-fluoropyridin-3-yl)carbonyl]morpholine
Catalog # APOSPC32691-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-[(2,6-Dichloro-5-fluoropyridin-3-yl)carbonyl]morpholine
Catalog # APOSPC32691-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :279.095 g/mol
Melting Pt:132…133 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:680217-88-3
Formula:C₁₀H₉Cl₂FN₂O₂
MDL Number:MFCD01765490