Order Entry
Italy
ContactUsLinkComponent
Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate 95%
Irritant
Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate 95%
Catalog # APOSPC3211-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate 95%
Catalog # APOSPC3211-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :266.243 g/mol
Boiling Pt:116…118 °C (0.5 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:175135-71-4
Formula:C₁₃H₁₂F₂N₂O₂
MDL Number:MFCD00067830