Order Entry
Italy
ContactUsLinkComponent
6-Chloro-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine
6-Chloro-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine
Catalog # APOSPC32092-500MG
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine
Catalog # APOSPC32092-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :298.651 g/mol
Storage Temperature:Ambient
CAS Number:175204-95-2
Formula:C₁₂H₆ClF₃N₄
MDL Number:MFCD00277273