
N-[(1,3-Benzodioxol-5-yl)methyl]-4-chloro-N-(4-fluorophenyl)-3-nitrobenzenesulphonamide
Catalog # APOSPC31161-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:464.851 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:216501-69-8
Formula:C₂₀H₁₄ClFN₂O₆S
MDL Number:MFCD00098183