3-Bromo-5,6-difluoro-2-methoxyphenylacetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1892087-78-3
:
Formula:C₉H₆BrF₂NO
MDL Number:MFCD32640084
:262.051 g/mol
Storage Temperature:Ambient
Frequently Bought Together