Order Entry
Italy
ContactUsLinkComponent
3-Bromo-2,6-difluorobenzamide 97%
3-Bromo-2,6-difluorobenzamide 97%
Catalog # APOSPC303451-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-2,6-difluorobenzamide 97%
Catalog # APOSPC303451-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.014 g/mol
Storage Temperature:Ambient
CAS Number:1541810-83-6
Formula:C₇H₄BrF₂NO
MDL Number:MFCD26680221