Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2-fluoro-5-methylbenzenesulphonamide
4-Chloro-2-fluoro-5-methylbenzenesulphonamide
4-Chloro-2-fluoro-5-methylbenzenesulphonamide
APOSPC303330-1G
:
Apollo Scientific
:
4-Chloro-2-fluoro-5-methylbenzenesulphonamide
APOSPC303330-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
223.652 g/mol
:
Ambient
:
1805525-24-9
:
C₇H₇ClFNO₂S
:
MFCD28751802