Order Entry
Italy
ContactUsLinkComponent
3,6-Difluoro-2-methoxyphenylacetonitrile 97%
3,6-Difluoro-2-methoxyphenylacetonitrile 97%
Catalog # APOSPC303316-1G
Supplier:  Apollo Scientific
CAS Number:  
3,6-Difluoro-2-methoxyphenylacetonitrile 97%
Catalog # APOSPC303316-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.155 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
CAS Number:1261760-73-9
Formula:C₉H₇F₂NO
MDL Number:MFCD18395261