Order Entry
Italy
ContactUsLinkComponent
4-Chloro-2,6-difluorobenzenesulphonyl chloride
4-Chloro-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC303234-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC303234-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.047 g/mol
Storage Temperature:Ambient
CAS Number:1208077-94-4
Formula:C₆H₂Cl₂F₂O₂S
MDL Number:MFCD12922627