Order Entry
Italy
ContactUsLinkComponent
2,4-Difluoro-3-methoxybenzenesulphonyl chloride
2,4-Difluoro-3-methoxybenzenesulphonyl chloride
Catalog # APOSPC303227-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Difluoro-3-methoxybenzenesulphonyl chloride
Catalog # APOSPC303227-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.628 g/mol
Storage Temperature:Ambient
CAS Number:1706458-65-2
Formula:C₇H₅ClF₂O₃S
MDL Number:MFCD28054212