2-Fluoro-4-methyl-3-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1824274-63-6
:
Formula:C₁₀H₈F₄O
MDL Number:MFCD28103360
:220.164 g/mol
Storage Temperature:Ambient
Frequently Bought Together