4-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde
Catalog # APOSPC303149-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.137 g/mol
Storage Temperature:Ambient
CAS Number:1706438-84-7
Formula:C₉H₆F₄O
MDL Number:MFCD28054194