2-Methoxy-5-(pentafluorosulphur)phenylacetic acid
Catalog # APOSPC303125-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:292.223 g/mol
Storage Temperature:Ambient
CAS Number:1240257-85-5
Formula:C₉H₉F₅O₃S
MDL Number:MFCD16652490