Order Entry
Italy
ContactUsLinkComponent
3-Chloro-4-ethoxy-5-fluorocinnamic acid 97%
3-Chloro-4-ethoxy-5-fluorocinnamic acid 97%
Catalog # APOSPC302946-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-4-ethoxy-5-fluorocinnamic acid 97%
Catalog # APOSPC302946-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.647 g/mol
Storage Temperature:Ambient
CAS Number:1017779-08-6
Formula:C₁₁H₁₀ClFO₃
MDL Number:MFCD09832348