
2-Fluoro-4-(trifluoromethyl)-DL-phenylalanine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:439587-15-2
:
Formula:C₁₀H₉F₄NO₂
MDL Number:MFCD04115995
:251.178 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together