2-Methoxy-5-(trifluoromethoxy)phenylacetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:773109-13-0
:
Formula:C₁₀H₉F₃O₄
MDL Number:MFCD04972913
:250.171 g/mol
Storage Temperature:Ambient
Frequently Bought Together