Ethyl 8-bromo-6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2514942-02-8
:
Formula:C₁₁H₈BrClF₂N₂O₂
Melting Pt:130 °C
:353.547 g/mol
Storage Temperature:Ambient
Frequently Bought Together