6-Bromo-2-(difluoromethyl)-8-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2451256-46-3
:
Formula:C₁₀H₇BrF₂N₂O₂
:305.076 g/mol
Melting Pt:207…209 °C
Storage Temperature:Ambient
Frequently Bought Together