Ethyl 6-bromo-2-(difluoromethyl)-8-methylimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2451256-39-4
:
Formula:C₁₂H₁₁BrF₂N₂O₂
:333.129 g/mol
Melting Pt:124…127 °C
Storage Temperature:Ambient
Frequently Bought Together