Ethyl (2-fluoro-5-methylphenoxy)acetate
Catalog # APOSPC301022-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.218 g/mol
Storage Temperature:Ambient
CAS Number:1339681-64-9
Formula:C₁₁H₁₃FO₃
MDL Number:MFCD18911516