
Ethyl 6-bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1121051-30-6
:
Formula:C₁₁H₈BrF₃N₂O₂
MDL Number:MFCD19704051
:337.093 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together