Order Entry
Italy
ContactUsLinkComponent
5-Fluoro-3-(4-methoxybenzylidene)-1,3-dihydro-2H-indol-2-one
5-Fluoro-3-(4-methoxybenzylidene)-1,3-dihydro-2H-indol-2-one
Catalog # APOSPC300930-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Fluoro-3-(4-methoxybenzylidene)-1,3-dihydro-2H-indol-2-one
Catalog # APOSPC300930-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.27 g/mol
Storage Temperature:Ambient
CAS Number:686726-22-7
Formula:C₁₆H₁₂FNO₂
MDL Number:MFCD14721317