Order Entry
Italy
ContactUsLinkComponent
Methyl 2-(3-chloro-4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate
Methyl 2-(3-chloro-4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate
Catalog # APOSPC300894-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-(3-chloro-4-fluorophenyl)imidazo[1,2-a]pyridine-6-carboxylate
Catalog # APOSPC300894-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :304.704 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀ClFN₂O₂
MDL Number:MFCD29918593