
1,1,1-Trifluoro-3-(1H-pyrazol-1-yl)propan-2-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1342051-08-4
:
Formula:C₆H₇F₃N₂O
MDL Number:MFCD19602661
:180.128 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together