2,5-Difluoro-4-morpholin-4-ylbenzoic acid
Catalog # APOSPC300787-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.207 g/mol
Storage Temperature:Ambient
CAS Number:1430223-92-9
Formula:C₁₁H₁₁F₂NO₃
MDL Number:MFCD28103348