Methyl 2,3,5-trifluoro-4-(1H-pyrazol-1-yl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1858250-92-6
:
Formula:C₁₁H₇F₃N₂O₂
MDL Number:MFCD28103346
:256.181 g/mol
Storage Temperature:Ambient
Frequently Bought Together