Order Entry
Italy
ContactUsLinkComponent
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl
Corrosive
Irritant
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl
Catalog # APOSPC300618-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl
Catalog # APOSPC300618-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.121 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:147497-57-2
Formula:C₁₃H₁₀BrF
MDL Number:MFCD15146810