Order Entry
Italy
ContactUsLinkComponent
7-Fluoro-6-methoxy-1-methyl-1H-indazol-3-amine
Irritant
7-Fluoro-6-methoxy-1-methyl-1H-indazol-3-amine
Catalog # APOSPC300573-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Fluoro-6-methoxy-1-methyl-1H-indazol-3-amine
Catalog # APOSPC300573-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.194 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1355171-72-0
Formula:C₉H₁₀FN₃O
MDL Number:MFCD16631266