
6-Bromo-8-(trifluoromethyl)imidazo[1,2-a]pyridine
Catalog # APOSPC300501-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:265.03 g/mol
Melting Pt:110…112 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1417334-55-4
Formula:C₈H₄BrF₃N₂
MDL Number:MFCD22577291