Order Entry
Italy
ContactUsLinkComponent
3-Fluoroprop-1-yl trifluoromethanesulphonate 95%
Flammable
Corrosive
3-Fluoroprop-1-yl trifluoromethanesulphonate 95%
Catalog # APOSPC2905-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
3-Fluoroprop-1-yl trifluoromethanesulphonate 95%
Catalog # APOSPC2905-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
ADR:8+3,II
 :210.147 g/mol
UN:2920
 :GHS02
 :GHS05
Storage Temperature:Ambient
CAS Number:180597-96-0
Formula:C₄H₆F₄O₃S
MDL Number:MFCD07784315